missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

cis-2,3-Epoxybutane, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1032006

 View more versions of this product

Catalog No. AAL1032006


Explore available promotions

Description

Description

Used in the enantioselective methanolysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the enantioselective methanolysis.

Solubility
Soluble in water, 2.848e+004 mg/L.

Notes
Moisture Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture. Store at 4°C.
Specifications

Specifications

cis-2,3-Epoxybutane
-84°C
60°C to 61°C
C4H8O
MFCD00005128
UN3271
cis-2,3-epoxybutane, cis-2-butene oxide, cis-2,3-dimethyloxirane, oxirane, 2,3-dimethyl-, cis, cis-2-butene epoxide, cis-2-butylene oxide, butane, 2,3-epoxy-, cis, ccris 3758, 2r,3s-2,3-dimethyloxirane, meso-2,3-epoxybutane
PQXKWPLDPFFDJP-ZXZARUISSA-N
(2R,3S)-2,3-dimethyloxirane
92162
98%
1758-33-4
0.826
−21°C (−5°F)
1.383
5 g
79770
Soluble in water,2.848e+004mg/L.
C[C@H]1O[C@H]1C
72.11
72.11

Chemical Identifiers

1758-33-4
72.11
PQXKWPLDPFFDJP-ZXZARUISSA-N
92162
C[C@H]1O[C@H]1C
C4H8O
MFCD00005128
cis-2,3-epoxybutane, cis-2-butene oxide, cis-2,3-dimethyloxirane, oxirane, 2,3-dimethyl-, cis, cis-2-butene epoxide, cis-2-butylene oxide, butane, 2,3-epoxy-, cis, ccris 3758, 2r,3s-2,3-dimethyloxirane, meso-2,3-epoxybutane
(2R,3S)-2,3-dimethyloxirane
Safety and Handling

Safety and Handling

GHS H Statement
H225-H341-H315-H319-H335
Highly flammable liquid and vapor.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c

H225-H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.

EINECSNumber : 000-000-0

RTECSNumber : EK3855020

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only