missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

cis-2-Butene-1,4-diol, 96%, remainder trans-isomer, Thermo Scientific Chemicals

$96.25 - $233.75

Chemical Identifiers

CAS 6117-80-2
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00002924,MFCD00063207
InChI Key ORTVZLZNOYNASJ-UPHRSURJSA-N
Synonym cis-2-butene-1,4-diol, z-2-butene-1,4-diol, cis-butenediol, unii-za7vgu6scv, cis-1,4-dihydroxy-2-butene, z-but-2-ene-1,4-diol, 2-butene-1,4-diol, 2z-but-2-ene-1,4-diol, 2-butene-1,4-diol, z, za7vgu6scv
PubChem CID 643790
IUPAC Name (Z)-but-2-ene-1,4-diol
SMILES OC\C=C/CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1918230
View Documents
Thermo Scientific Chemicals
A1918230
250 g
Each for $96.25
 
AAA191820B
View Documents
Thermo Scientific Chemicals
A191820B
1000 g
Each for $233.75
 
Description

Description

cis-2-Butene-1, 4-diol is used as a probe for studying isomerization versus hydrogenation and hydrogenolysis reactions. (DPCB-OMe) efficiently catalyzes cyclodehydration of cis-2-butene-1,4-diol with active methylene compounds to give 2-vinyl-2,3-dihydrofurans in good to high yields. Grubbs's cross metathesis of eugenol with cis-2-butene-1, 4-diol to make a natural product, an organometallic experiment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-2-Butene-1, 4-diol is used as a probe for studying isomerization versus hydrogenation and hydrogenolysis reactions. (DPCB-OMe) efficiently catalyzes cyclodehydration of cis-2-butene-1,4-diol with active methylene compounds to give 2-vinyl-2,3-dihydrofurans in good to high yields. Grubbs′s cross metathesis of eugenol with cis-2-butene-1, 4-diol to make a natural product, an organometallic experiment.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

6117-80-2
88.11
ORTVZLZNOYNASJ-UPHRSURJSA-N
643790
OC\C=C/CO
C4H8O2
MFCD00002924,MFCD00063207
cis-2-butene-1,4-diol, z-2-butene-1,4-diol, cis-butenediol, unii-za7vgu6scv, cis-1,4-dihydroxy-2-butene, z-but-2-ene-1,4-diol, 2-butene-1,4-diol, 2z-but-2-ene-1,4-diol, 2-butene-1,4-diol, z, za7vgu6scv
(Z)-but-2-ene-1,4-diol

Specifications

6117-80-2
1.07
127°C (260°F)
1.478
250 g
cis-2-butene-1,4-diol, z-2-butene-1,4-diol, cis-butenediol, unii-za7vgu6scv, cis-1,4-dihydroxy-2-butene, z-but-2-ene-1,4-diol, 2-butene-1,4-diol, 2z-but-2-ene-1,4-diol, 2-butene-1,4-diol, z, za7vgu6scv
ORTVZLZNOYNASJ-UPHRSURJSA-N
(Z)-but-2-ene-1,4-diol
643790
96%
remainder trans-isomer
6°C to 8°C
131°C to 132°C (12 mmHg)
C4H8O2
MFCD00002924,MFCD00063207
1679241
Soluble in water.
OC\C=C/CO
88.11
88.11
cis-2-Butene-1,4-diol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 228-085-1

RTECSNumber : EM4970000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only