cis-2-Heptene 95.0+%, TCI America™
Supplier:  TCI America H041110ML
 View more versions of this product
Catalog No. H041110ML
Explore available promotions
Description
Description
Specifications
Specifications
cis-2-Heptene
Colorless
C7H14
10 mL
OTTZHAVKAVGASB-HYXAFXHYSA-N
(Z)-hept-2-ene
643836
≥95.0% (GC)
6443-92-1
99°C
MFCD00063976
3295
CCCCC=CC
98.189
98.19
Liquid
Chemical Identifiers
6443-92-1
98.189
OTTZHAVKAVGASB-HYXAFXHYSA-N
(Z)-hept-2-ene
C7H14
MFCD00063976
643836
CCCCC=CC
Videos
Safety and Handling
Safety and Handling
TSCA : No