Learn More
cis-4-(Boc-amino)cyclohexanemethylamine, 97%
CAS: 509143-00-4 | C12H24N2O2 | 228.336 g/mol
$233.01 - $752.01
Chemical Identifiers
| CAS | 509143-00-4 |
|---|---|
| Molecular Formula | C12H24N2O2 |
| Molecular Weight (g/mol) | 228.336 |
| MDL Number | MFCD03844602 |
| InChI Key | NVQFOBONHIXDOC-UHFFFAOYSA-N |
| Synonym | tert-butyl trans-4-aminomethyl cyclohexyl carbamate, tert-butyl n-4-aminomethyl cyclohexyl carbamate, tert-butyl cis-4-aminomethylcyclohexylcarbamate, tert-butyl n-1r,4r-4-aminomethyl cyclohexyl carbamate, tert-butyl trans-4-aminomethylcyclohexyl carbamate, tert-butyl cis-4-aminomethyl cyclohexyl carbamate, cis-4-boc-amino cyclohexylmethylamine, tert-butyl trans-4-aminomethylcyclohexylcarbamate, trans-4-boc-amino cyclohexylmethylamine, tert-butyl 4-aminomethyl cyclohexyl carbamate |
| PubChem CID | 2756045 |
| IUPAC Name | tert-butyl N-[4-(aminomethyl)cyclohexyl]carbamate |
| SMILES | CC(C)(C)OC(=O)NC1CCC(CC1)CN |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH62187MD
|
Thermo Scientific Chemicals
H62187MD |
250 mg |
Each for $233.01
|
|
|||||
|
AAH6218703
|
Thermo Scientific Chemicals
H6218703 |
1 g |
Each for $752.01
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
NotesStore away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
| 509143-00-4 | |
| 228.336 | |
| NVQFOBONHIXDOC-UHFFFAOYSA-N | |
| 2756045 | |
| CC(C)(C)OC(=O)NC1CCC(CC1)CN |
| C12H24N2O2 | |
| MFCD03844602 | |
| tert-butyl trans-4-aminomethyl cyclohexyl carbamate, tert-butyl n-4-aminomethyl cyclohexyl carbamate, tert-butyl cis-4-aminomethylcyclohexylcarbamate, tert-butyl n-1r,4r-4-aminomethyl cyclohexyl carbamate, tert-butyl trans-4-aminomethylcyclohexyl carbamate, tert-butyl cis-4-aminomethyl cyclohexyl carbamate, cis-4-boc-amino cyclohexylmethylamine, tert-butyl trans-4-aminomethylcyclohexylcarbamate, trans-4-boc-amino cyclohexylmethylamine, tert-butyl 4-aminomethyl cyclohexyl carbamate | |
| tert-butyl N-[4-(aminomethyl)cyclohexyl]carbamate |
Specifications
| 509143-00-4 | |
| MFCD03844602 | |
| NVQFOBONHIXDOC-UHFFFAOYSA-N | |
| tert-butyl N-[4-(aminomethyl)cyclohexyl]carbamate | |
| 228.336 | |
| 228.33 | |
| cis-4-(Boc-amino)cyclohexanemethylamine |
| C12H24N2O2 | |
| tert-butyl trans-4-aminomethyl cyclohexyl carbamate, tert-butyl n-4-aminomethyl cyclohexyl carbamate, tert-butyl cis-4-aminomethylcyclohexylcarbamate, tert-butyl n-1r,4r-4-aminomethyl cyclohexyl carbamate, tert-butyl trans-4-aminomethylcyclohexyl carbamate, tert-butyl cis-4-aminomethyl cyclohexyl carbamate, cis-4-boc-amino cyclohexylmethylamine, tert-butyl trans-4-aminomethylcyclohexylcarbamate, trans-4-boc-amino cyclohexylmethylamine, tert-butyl 4-aminomethyl cyclohexyl carbamate | |
| CC(C)(C)OC(=O)NC1CCC(CC1)CN | |
| 250 mg | |
| 2756045 | |
| 97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only