missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

cis-4-Cyclohexene-1,2-dicarboxylic acid, 98%, Thermo Scientific Chemicals

$103.92 - $273.93

Chemical Identifiers

CAS 2305-26-2
Molecular Formula C8H10O4
Molecular Weight (g/mol) 170.164
MDL Number MFCD00070482
InChI Key ILUAAIDVFMVTAU-OLQVQODUSA-N
Synonym cis-4-cyclohexene-1,2-dicarboxylic acid, 1r,2s-rel-cyclohex-4-ene-1,2-dicarboxylic acid, 1r,2s-cyclohex-4-ene-1,2-dicarboxylic acid, 4-cyclohexene-1,2-dicarboxylic acid, cis, cis-delta4-tetrahydrophthalic acid, 4-cyclohexene-1,2-dicarboxylic acid, 1r,2s-rel, rel-1r,2s-cyclohex-4-ene-1,2-dicarboxylic acid, 1r,2s-rel-4-cyclohexene-1,2-dicarboxylic acid, cis-.delta.4-tetrahydrophthalic acid, cis-.delta.4-cyclohexene-1,2-dicarboxylic acid
PubChem CID 504243
IUPAC Name (1S,2R)-cyclohex-4-ene-1,2-dicarboxylic acid
SMILES C1C=CCC(C1C(=O)O)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0939214
View Documents
Thermo Scientific Chemicals
L0939214
25 g
Each for $103.92
 
AAL0939222
View Documents
Thermo Scientific Chemicals
L0939222
100 g
Each for $273.93
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2305-26-2
170.164
ILUAAIDVFMVTAU-OLQVQODUSA-N
504243
C1C=CCC(C1C(=O)O)C(=O)O
C8H10O4
MFCD00070482
cis-4-cyclohexene-1,2-dicarboxylic acid, 1r,2s-rel-cyclohex-4-ene-1,2-dicarboxylic acid, 1r,2s-cyclohex-4-ene-1,2-dicarboxylic acid, 4-cyclohexene-1,2-dicarboxylic acid, cis, cis-delta4-tetrahydrophthalic acid, 4-cyclohexene-1,2-dicarboxylic acid, 1r,2s-rel, rel-1r,2s-cyclohex-4-ene-1,2-dicarboxylic acid, 1r,2s-rel-4-cyclohexene-1,2-dicarboxylic acid, cis-.delta.4-tetrahydrophthalic acid, cis-.delta.4-cyclohexene-1,2-dicarboxylic acid
(1S,2R)-cyclohex-4-ene-1,2-dicarboxylic acid

Specifications

2305-26-2
C8H10O4
25 g
cis-4-cyclohexene-1,2-dicarboxylic acid, 1r,2s-rel-cyclohex-4-ene-1,2-dicarboxylic acid, 1r,2s-cyclohex-4-ene-1,2-dicarboxylic acid, 4-cyclohexene-1,2-dicarboxylic acid, cis, cis-delta4-tetrahydrophthalic acid, 4-cyclohexene-1,2-dicarboxylic acid, 1r,2s-rel, rel-1r,2s-cyclohex-4-ene-1,2-dicarboxylic acid, 1r,2s-rel-4-cyclohexene-1,2-dicarboxylic acid, cis-.delta.4-tetrahydrophthalic acid, cis-.delta.4-cyclohexene-1,2-dicarboxylic acid
C1C=CCC(C1C(=O)O)C(=O)O
170.164
170.17
cis-4-Cyclohexene-1,2-dicarboxylic acid
165°C to 170°C
MFCD00070482
2050419
ILUAAIDVFMVTAU-OLQVQODUSA-N
(1S,2R)-cyclohex-4-ene-1,2-dicarboxylic acid
504243
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 218-974-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only