missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(+/-)-cis-6-Hydroxy-1-methylbicyclo[4.3.0]nonane-2,7-dione 98.0+%, TCI America™
Supplier: TCI America H131850MG
Specifications
(+/-)-cis-6-Hydroxy-1-methylbicyclo[4.3.0]nonane-2,7-dione | |
101°C | |
50 mg | |
CAYWOGVLFCOLCK-VHSXEESVSA-N | |
(3aR,7aS)-7a-hydroxy-3a-methyl-3,5,6,7-tetrahydro-2H-indene-1,4-dione | |
50996020 | |
≥98.0% (GC) |
1192178-33-8 | |
C10H14O3 | |
(+/-)-cis-Hexahydro-7a-hydroxy-3a-methyl-1H-indene-1,4(2H)-dione | |
CC12CCC(=O)C1(CCCC2=O)O | |
182.219 | |
182.22 | |
Crystalline Powder |
Chemical Identifiers
1192178-33-8 | |
182.219 | |
(+/-)-cis-Hexahydro-7a-hydroxy-3a-methyl-1H-indene-1,4(2H)-dione | |
(3aR,7aS)-7a-hydroxy-3a-methyl-3,5,6,7-tetrahydro-2H-indene-1,4-dione |
C10H14O3 | |
CAYWOGVLFCOLCK-VHSXEESVSA-N | |
50996020 | |
CC12CCC(=O)C1(CCCC2=O)O |
Safety and Handling
TSCA : No