Learn More
cis-Cyclooctene, 95%, stab., Thermo Scientific Chemicals
$82.73 - $290.36
Chemical Identifiers
CAS | 931-87-3 |
---|---|
Molecular Formula | C8H14 |
Molecular Weight (g/mol) | 110.20 |
MDL Number | MFCD00001753 |
InChI Key | URYYVOIYTNXXBN-UPHRSURJSA-N |
Synonym | cis-cyclooctene, z-cyclooctene, cyclooctene, z, cyclooctene, 1z, cyclooct-1-ene, e-cyclooctene, 1-cyclooctene #, 1z-cyclooctene, 7z-cyclooctene, ar,e-cyclooctene |
PubChem CID | 638079 |
SMILES | C1CCC\C=C/CC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA13477AD
|
Thermo Scientific Chemicals
A13477AD |
50 mL |
Each for $82.73
|
|
||||||
AAA13477AE
|
Thermo Scientific Chemicals
A13477AE |
100 mL |
Each for $98.78
|
|
||||||
AAA13477AK
|
Thermo Scientific Chemicals
A13477AK |
250 mL |
Each for $164.90
|
|
||||||
AAA13477AP
|
Thermo Scientific Chemicals
A13477AP |
500 mL |
Each for $290.36
|
|
||||||
Description
cis-Cyclooctene is used as a displaced ligand in chlorobis(cyclooctene)rhodium dimer and chlorobis(cyclooctene)iridium dimer in organometallic chemistry. It acts as a monomer used in synthetic chemistry. It is used to prepare 1-chloro-4-(trichloromethyl) cyclooctane by reaction with carbon tetrachloride using dichlorotris(triphenylphosphine) ruthenium(II) as a catalyst. Further, it is used to study the alkenes chemisorbed on silicon(100) by scanning tunneling microscopy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationscis-Cyclooctene is used as a displaced ligand in chlorobis(cyclooctene)rhodium dimer and chlorobis(cyclooctene)iridium dimer in organometallic chemistry. It acts as a monomer used in synthetic chemistry. It is used to prepare 1-chloro-4-(trichloromethyl) cyclooctane by reaction with carbon tetrachloride using dichlorotris(triphenylphosphine) ruthenium(II) as a catalyst. Further, it is used to study the alkenes chemisorbed on silicon(100) by scanning tunneling microscopy.
Solubility
Miscible with alcohol and ether. Immiscible with water.
Notes
Air sensitive. Incompatible with strong oxidizing agents.
Chemical Identifiers
931-87-3 | |
110.20 | |
URYYVOIYTNXXBN-UPHRSURJSA-N | |
638079 |
C8H14 | |
MFCD00001753 | |
cis-cyclooctene, z-cyclooctene, cyclooctene, z, cyclooctene, 1z, cyclooct-1-ene, e-cyclooctene, 1-cyclooctene #, 1z-cyclooctene, 7z-cyclooctene, ar,e-cyclooctene | |
C1CCC\C=C/CC1 |
Specifications
931-87-3 | |
0.848 | |
25°C (77°F) | |
C8H14 | |
MFCD00001753 | |
UN3295 | |
Air Sensitive | |
Miscible with alcohol and ether. Immiscible with water. | |
C1CCC\C=C/CC1 | |
110.20 | |
110.2 | |
cis-Cyclooctene, stabilized |
-16°C | |
145°C to 146°C | |
Recognizable | |
1.47 | |
50 mL | |
1280166 | |
cis-cyclooctene, z-cyclooctene, cyclooctene, z, cyclooctene, 1z, cyclooct-1-ene, e-cyclooctene, 1-cyclooctene #, 1z-cyclooctene, 7z-cyclooctene, ar,e-cyclooctene | |
URYYVOIYTNXXBN-UPHRSURJSA-N | |
(Z)-cyclooctene | |
638079 | |
95% |
Safety and Handling
GHS H Statement
H304-H226
May be fatal if swallowed and enters airways.
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
H226-H304
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber : 213-243-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only