Learn More
cis-Dichlorobis(diethylsulfide)platinum(II), Pt 43.0% min, Thermo Scientific Chemicals
Hydrogenation
$138.96 - $224.62
Chemical Identifiers
CAS | 15442-57-6 |
---|---|
Molecular Formula | C8H20Cl2PtS2 |
Molecular Weight (g/mol) | 446.35 |
MDL Number | MFCD00050943 |
InChI Key | RFSLRYGKHBDFGC-UHFFFAOYSA-L |
Synonym | cis-dichlorobis diethylsulfide platinum ii, cis-dichlorobis diethyl sulfide platinum ii, dichloroplatinum, ethylsulfanylethane, trans-dichlorobis diethylsulfide platinum ii, cis-bis diethyl sulfide platinum ii chloride, dichlorobis 1,1'-thiobis ethane platinum, platinum, dichlorobis 1,1'-thiobis ethane, dichloroplatinum; ethylsulfanylethane, ethylsulfanyl ethane-dichloroplatinum 2:1 |
PubChem CID | 167183 |
SMILES | Cl[Pt++]Cl.CCSCC.CCSCC |
Description
Hydrogenation
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
15442-57-6 | |
446.35 | |
RFSLRYGKHBDFGC-UHFFFAOYSA-L | |
167183 |
C8H20Cl2PtS2 | |
MFCD00050943 | |
cis-dichlorobis diethylsulfide platinum ii, cis-dichlorobis diethyl sulfide platinum ii, dichloroplatinum, ethylsulfanylethane, trans-dichlorobis diethylsulfide platinum ii, cis-bis diethyl sulfide platinum ii chloride, dichlorobis 1,1'-thiobis ethane platinum, platinum, dichlorobis 1,1'-thiobis ethane, dichloroplatinum; ethylsulfanylethane, ethylsulfanyl ethane-dichloroplatinum 2:1 | |
Cl[Pt++]Cl.CCSCC.CCSCC |
Specifications
15442-57-6 | |
MFCD00050943 | |
cis-dichlorobis diethylsulfide platinum ii, cis-dichlorobis diethyl sulfide platinum ii, dichloroplatinum, ethylsulfanylethane, trans-dichlorobis diethylsulfide platinum ii, cis-bis diethyl sulfide platinum ii chloride, dichlorobis 1,1'-thiobis ethane platinum, platinum, dichlorobis 1,1'-thiobis ethane, dichloroplatinum; ethylsulfanylethane, ethylsulfanyl ethane-dichloroplatinum 2:1 | |
RFSLRYGKHBDFGC-UHFFFAOYSA-L | |
dichloroplatinum;ethylsulfanylethane | |
167183 | |
Pt 43.7% | |
cis-Dichlorobis(diethylsulfide)platinum(II) |
C8H20Cl2PtS2 | |
0.5 g | |
Soluble in acetone, alcohol and benzene | |
Cl[Pt++]Cl.CCSCC.CCSCC | |
446.35 | |
446.37 | |
Crystalline |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 239-454-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only