missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

cis-Dichlorobis(triethylphosphine)platinum(II), Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals 039284ME

Catalog No. AA39284ME


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

cis-Dichlorobis(triethylphosphine)platinum(II)
139°C to 142°C
MFCD00058821
Hygroscopic
WPWLTKRUFHHDLP-UHFFFAOYSA-L
dichloroplatinum;triethylphosphane
11027520
Powder
15692-07-6
C12H30Cl2P2Pt
500 mg
cis-dichlorobis triethylphosphine platinum ii, et3p 2ptcl2, trans-dichlorobis triethylphosphine platinum ii, cis-bis triethylphosphine platinum dichloride, bis triethylphosphine platinum dichloride, cis-bis triethylphosphine platinum cento chloride, dichlorobis triethylphosphine platinum ii, cis-bis triethylphosphine platinum ii chloride
[Cl-].[Cl-].[Pt++].CCP(CC)CC.CCP(CC)CC
502.30
502.32

Chemical Identifiers

15692-07-6
502.30
WPWLTKRUFHHDLP-UHFFFAOYSA-L
11027520
[Cl-].[Cl-].[Pt++].CCP(CC)CC.CCP(CC)CC
C12H30Cl2P2Pt
MFCD00058821
cis-dichlorobis triethylphosphine platinum ii, et3p 2ptcl2, trans-dichlorobis triethylphosphine platinum ii, cis-bis triethylphosphine platinum dichloride, bis triethylphosphine platinum dichloride, cis-bis triethylphosphine platinum cento chloride, dichlorobis triethylphosphine platinum ii, cis-bis triethylphosphine platinum ii chloride
dichloroplatinum;triethylphosphane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c

H315-H317-H319-H334-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only