missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Citric Acid, 10% (w/v) from USP grade material, Ricca Chemical

Supplier:  Ricca Chemical Company 212016

 View more versions of this product

Catalog No. 212016


Explore available promotions

Description

Description

Specifications

Specifications

Citric Acid
89.0,9.0
93.0,11.0
MFCD00011669
KRKNYBCHXYNGOX-UHFFFAOYSA-N
10% (w/v)
192.12
CHEBI:30769
Natural Poly Bottle
77-92-9,7732-18-5
Colorless
C6H8O7
citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
OC(=O)CC(O)(CC(O)=O)C(O)=O
2-hydroxypropane-1,2,3-tricarboxylic acid
311
500 mL
Liquid

Chemical Identifiers

77-92-9
192.12
KRKNYBCHXYNGOX-UHFFFAOYSA-N
311
2-hydroxypropane-1,2,3-tricarboxylic acid
C6H8O7
MFCD00011669
citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
CHEBI:30769
OC(=O)CC(O)(CC(O)=O)C(O)=O
SDS
Documents

Documents

Promotions

Promotions