missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Citric Acid, Anhydrous p.a., Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 220345000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer for analysisSpecifications
Citric acid | |
77-92-9 | |
99.5% | |
HO2CCH2C(OH)(CO2H)CH2CO2H | |
citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy | |
KRKNYBCHXYNGOX-UHFFFAOYSA-N | |
2-hydroxypropane-1,2,3-tricarboxylic acid | |
311 | |
192.13 | |
Analytical | |
100°C | |
0.5% max. | |
Crystals or Crystalline Powder |
153.0°C | |
White | |
C6H8O7 | |
MFCD00011669 | |
Solubility in water: 750g/L (20°C). Other solubilities: soluble in alcohol and ether, badly soluble in diethyl ether, insoluble in benzene and chloroform | |
OC(=O)CC(O)(CC(O)=O)C(O)=O | |
192.12 | |
CHEBI:30769 | |
99.5% | |
500 g | |
Authentic | |
Plastic bottle |
Chemical Identifiers
77-92-9 | |
192.12 | |
KRKNYBCHXYNGOX-UHFFFAOYSA-N | |
311 | |
2-hydroxypropane-1,2,3-tricarboxylic acid |
C6H8O7 | |
MFCD00011669 | |
citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy | |
CHEBI:30769 | |
OC(=O)CC(O)(CC(O)=O)C(O)=O |
Safety and Handling
Warning
RUO – Research Use Only