missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Citric acid, diammonium salt, ACS reagent, Thermo Scientific Chemicals
$242.11 - $1298.05
Chemical Identifiers
CAS | 3012-65-5 |
---|---|
Molecular Formula | C6H14N2O7 |
Molecular Weight (g/mol) | 226.19 |
MDL Number | MFCD00013068 |
InChI Key | YXVFQADLFFNVDS-UHFFFAOYSA-N |
Synonym | ammonium citrate dibasic, ammonium hydrogencitrate, diammonium hydrogen citrate, citric acid diammonium salt, citric acid ammonium salt, di-ammonium hydrogen citrate, citro diamine, citric acid diamine, ammonium citrate, dibasic, acs, ammonium citrate dibasic, puriss |
PubChem CID | 13710713 |
ChEBI | CHEBI:63076 |
IUPAC Name | azane;2-hydroxypropane-1,2,3-tricarboxylic acid |
SMILES | [NH4+].[NH4+].OC(=O)C(O)(CC([O-])=O)CC([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC447685000
|
Thermo Scientific Chemicals
447685000 |
500 g |
Each for $242.11
|
|
|||||
AC447680025
|
Thermo Scientific Chemicals
447680025 |
2.5 kg |
Each for $1,298.05
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3012-65-5 | |
226.19 | |
YXVFQADLFFNVDS-UHFFFAOYSA-N | |
13710713 | |
azane;2-hydroxypropane-1,2,3-tricarboxylic acid |
C6H14N2O7 | |
MFCD00013068 | |
ammonium citrate dibasic, ammonium hydrogencitrate, diammonium hydrogen citrate, citric acid diammonium salt, citric acid ammonium salt, di-ammonium hydrogen citrate, citro diamine, citric acid diamine, ammonium citrate, dibasic, acs, ammonium citrate dibasic, puriss | |
CHEBI:63076 | |
[NH4+].[NH4+].OC(=O)C(O)(CC([O-])=O)CC([O-])=O |
Specifications
3012-65-5 | |
0.01% max | |
C6H14N2O7 | |
500 g | |
15509 | |
Solubility in water: soluble | |
[NH4+].[NH4+].OC(=O)C(O)(CC([O-])=O)CC([O-])=O | |
226.19 | |
CHEBI:63076 | |
ACS Reagent | |
Citric acid, diammonium salt |
1.5000g/mL | |
Plastic bottle | |
MFCD00013068 | |
1.5 | |
ammonium citrate dibasic, ammonium hydrogencitrate, diammonium hydrogen citrate, citric acid diammonium salt, citric acid ammonium salt, di-ammonium hydrogen citrate, citro diamine, citric acid diamine, ammonium citrate, dibasic, acs, ammonium citrate dibasic, puriss | |
YXVFQADLFFNVDS-UHFFFAOYSA-N | |
azane;2-hydroxypropane-1,2,3-tricarboxylic acid | |
13710713 | |
226.19 | |
0.005% max |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 221-146-3
RUO – Research Use Only