missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Citric Acid, Monohydrate, Granular, USP, 99.5-100.5%, Spectrum™ Chemical

CI132, 5949-29-1, C6H8O7·H2O

Supplier:  Spectrum Chemical Mfg Cor CI132500GM

 View more versions of this product

Catalog No. 18601729


Explore available promotions

Description

Description

Spectrum™ Chemical Citric Acid, Monohydrate, Granular, USP is used as thrombin inhibitor and fungicide in pharmaceutical prepartions. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Specifications

5949-29-1
C6H10O8
O.OC(=O)CC(O)(CC(O)=O)C(O)=O
210.14
USP
0.1%
100%
YASYEJJMZJALEJ-UHFFFAOYSA-N
2-hydroxypropane-1,2,3-tricarboxylic acid hydrate
99.5 to 100.5%
500 g
Poly Bottle

Chemical Identifiers

5949-29-1
210.14
2-hydroxypropane-1,2,3-tricarboxylic acid hydrate
C6H10O8
YASYEJJMZJALEJ-UHFFFAOYSA-N
O.OC(=O)CC(O)(CC(O)=O)C(O)=O
Videos
SDS
Documents

Documents

Promotions

Promotions