missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Citric acid, trilithium salt tetrahydrate, 98%, pure, Thermo Scientific Chemicals

$281.48 - $281.48

Chemical Identifiers

CAS 6080-58-6
MDL Number MFCD00150030
InChI Key HXGWMCJZLNWEBC-UHFFFAOYSA-K
Synonym lithium citrate tetrahydrate, unii-5z6e9k79yv, trilithium citrate tetrahydrate, lithium citrate usp, citric acid trilithium salt tetrahydrate, tri-lithium citrate tetrahydrate, lithium citrate tribasic tetrahydrate, lithonate tn, 1,2,3-propanetricarboxylic acid, 2-hydroxy-trilithium salt tetrahydrate
PubChem CID 2724118
ChEBI CHEBI:64754
IUPAC Name trilithium;2-hydroxypropane-1,2,3-tricarboxylate;tetrahydrate
SMILES [Li+].[Li+].[Li+].C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC199855000
SDS View Documents
Thermo Scientific Chemicals
199855000
500 g Plastic bottle
Each for $281.48
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Chemical Identifiers

6080-58-6
HXGWMCJZLNWEBC-UHFFFAOYSA-K
2724118
trilithium;2-hydroxypropane-1,2,3-tricarboxylate;tetrahydrate
MFCD00150030
lithium citrate tetrahydrate, unii-5z6e9k79yv, trilithium citrate tetrahydrate, lithium citrate usp, citric acid trilithium salt tetrahydrate, tri-lithium citrate tetrahydrate, lithium citrate tribasic tetrahydrate, lithonate tn, 1,2,3-propanetricarboxylic acid, 2-hydroxy-trilithium salt tetrahydrate
CHEBI:64754
[Li+].[Li+].[Li+].C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.O

Specifications

6080-58-6
White
98%
C6H5Li3O7·4H2O
MFCD00150030
03, I, 196
lithium citrate tetrahydrate, unii-5z6e9k79yv, trilithium citrate tetrahydrate, lithium citrate usp, citric acid trilithium salt tetrahydrate, tri-lithium citrate tetrahydrate, lithium citrate tribasic tetrahydrate, lithonate tn, 1,2,3-propanetricarboxylic acid, 2-hydroxy-trilithium salt tetrahydrate
HXGWMCJZLNWEBC-UHFFFAOYSA-K
trilithium;2-hydroxypropane-1,2,3-tricarboxylate;tetrahydrate
2724118
281.97
Pure
Citric acid, trilithium salt tetrahydrate, 98%
112.0°C
Authentic
Plastic bottle
LiO2CCH2C(OH)(CO2Li)CH2CO2Li·4H2O
500 g
15, 5586
Solubility in water: 745g/L water (25°C). Other solubilities: slightly soluble in alcohol
[Li+].[Li+].[Li+].C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.O
281.97
CHEBI:64754
98%
Fine Crystalline Powder
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only