Thermo Scientific™ Clamp for 250mL Flask for Precision™ Water Baths
Use these flask clips and fasteners in Thermo Scientific™ Precision Reciprocal, Dubnoff and Shallow-Form Shaking Water Baths.
Supplier:  Thermo Scientific™ 3166566
Catalog No. 15-455-15E
Explore available promotions
Description
Description
Clips and fasteners for use with Thermo Scientific™ Precision water baths.
Flask Clips
  • Secure various size flasks to bath platform
  • Stainless steel
  • Each requires one fastener (Cat. No. 3166189 - order separately)
Test Tube Clip
  • Stainless steel
  • Hold 13mm to 25mm tubes
  • Each requires one fastener (Cat. No. 3166189 - order separately)