Eisco™ Class A Glass Volumetric Flasks with White Graduations
Eisco™ Class A Glass Volumetric Flasks with White Graduations feature a flat bottom design, interchangeable solid glass stoppers, and single graduation.
Supplier:  Eisco™ CH0441DWT
 View more versions of this product
Catalog No. S89398
Explore available promotions
Description
Description
  • Include solid glass stoppers
  • Flat bottom design
  • Single graduation mark
  • White printed markings
  • Class A, in compliance with ASTM-E288
  • Autoclavable
Warranty
1 year limited manufacturer warranty
Specifications
Specifications
Clear
Clear
Borosilicate Glass (3.3)
16.1 in.
6.9 in.
Long
±0.1 mL
5.5 cm
Reusable
Beaded
A
Glass Stopper
Stopper No. 16
Borosilicate Glass 3.3
200 mL
Prepare chemical solution
19 cm
5.5 cm
Volumetric Flask
6.9 in.
Autoclavable
Yes
Non-sterile
Flat
Non-treated