Eisco™ Class A Round Base Glass Graduated Cylinders
Eisco™ Class A Round Base Glass Graduated Cylinders feature printed graduations and spouts.
Supplier:  Eisco™ CH0344LWT
 View more versions of this product
Catalog No. S88438
Explore available promotions
Description
Description
  • Made from high quality Borosilicate 3.3 Glass
  • Single piece construction with round base
  • Printed graduations are chemical resistant
  • Spout for easy, mess free pouring
  • Class A
  • Conforms to DIN EN ISO 4788 standard
  • Autoclavable
Specifications
Specifications
Round
Graduated Measuring Cylinder
White
Round
25 cm
100 mL
1 mL
±0.5 mL
9.75 in.
DIN EN ISO 4788
A
Borosilicate Glass
Measuring Liquids
Autoclavable