Eisco™ Class B Glass Burettes with Glass Key Stopcock
Eisco™ Class B Glass Burettes with Glass Key Stopcock designed with blue graduations and a straight single bore.
Supplier:  Eisco™ CH0236H
 View more versions of this product
Catalog No. S29399
Explore available promotions
Description
Description
  • Made from high quality 3.3 borosilicate glass
  • Blue ceramic graduations
  • Straight single bore
  • Includes smooth glass key stopcock
  • Glass Key stopcock requires lubrication but offers the smoothest, most authentic control
Warranty
1 year limited manufacturer warranty
Specifications
Specifications
100 mL
Borosilicate Glass
Dispensing Liquid
Straight
±0.2 mL
Class B
2 cm
0.2 mL
77.5 cm
Glass
Straight Bore Buret