Humboldt Clay Pipe Iron-Wire Triangles
Supplier:  Humboldt H23920
 View more versions of this product
Catalog No. 15-285C
Explore available promotions
Description
Description
Crucibles completely exposed to flame. They rest on three flanges of clay pipe stems covering iron wire of triangle.
Specifications
Specifications
Iron Wire
7.62 cm
Flanged pipe stem
3 in.
16g
Triangle