MilliporeSigma™ Supelco™ Cleaned Copper Tubing, Length: 50 ft.
Solvent-washed to ASTM B-280 specifications
Supplier: MilliporeSigma™ Supelco™ 20488
Specifications
Solvent-Washed to ASTM B-280 Specifications | |
0.065 in. | |
1.65 mm | |
50 ft. |
GC Plumbing Applications | |
1/8 in. | |
3.18 mm | |
Copper |