PURUS™ CleanImage Premier Cleanroom Paper
For writing, printing and photocopying in cleanroom environments, PURUS™ Cleanroom Paper provides high printability and heat toner adhesion, and is also recyclable.
Supplier:  PURUS™ PCIB1084C
 View more versions of this product
Catalog No. 19-125-892
Explore available promotions
Description
Description
  • Propriatary Binding Technology
  • Suitable for use in ISO 4 to 6 (Class 10 to Class 1000) Cleanroom
  • Produced in ISO 4 (Class 10 Cleanroom)
  • Recyclable
  • High printability and heat toner adhesion
Compliance
Suitable for use in ISO 4 to 6 (Class 10 to Class 1000).
Specifications
Specifications
Polyethylene
11.7 in.
Blue
210 x 297 mm (8.3 x 11.7 in.)
Writing, Printing and Photocopying in Cleanroom Environments
Yes
250 Sheets/Pk., 10 Pk./Cs.
22 lb.
No
8.3 in.
Blue
Autoclavable
Holeless
Cleanroom Paper
A4
105 gsm