missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PURUS™ CleanImage Premier Cleanroom Paper

For writing, printing and photocopying in cleanroom environments, PURUS™ Cleanroom Paper provides high printability and heat toner adhesion, and is also recyclable.

Supplier:  PURUS™ PCIK1088C

 View more versions of this product

Catalog No. 19-125-895


Explore available promotions

Description

Description

  • Propriatary Binding Technology
  • Suitable for use in ISO 4 to 6 (Class 10 to Class 1000) Cleanroom
  • Produced in ISO 4 (Class 10 Cleanroom)
  • Recyclable
  • High printability and heat toner adhesion

Compliance

Suitable for use in ISO 4 to 6 (Class 10 to Class 1000).

Specifications

Specifications

Polyethylene
11 in.
Pink
216 x 279.4 mm (8.5 x 11 in.)
279 mm
216 mm
Laser Printers and Copy Machines
Cleanroom Paper
9.97 kg
No
8.5 in.
Pink
11 in.
8.5 in.
Writing, Printing and Photocopying in Cleanroom Environments
Holeless
22 lb.
Videos
SDS
Documents

Documents

Promotions

Promotions