Fisherbrand™ Cleanroom Notebooks, Latex
European sizing cleanroom notebooks, college ruled.
Supplier: Fisherbrand™ 18001500
Description
- For documentation in critical environments
- Low particulate and ESD generation
- Printed on #22 paper with college rule lines.
- Semi-transparent high density polyethylene (HDPE) covers
- 100 pages/50 sheets
- Spiral bound
Specifications
Latex | |
HDPE | |
8.5 x 11 in. |
College Ruled | |
20/case | |
ISO Clean 10 |