Fisherbrand™ Cleanroom Pens
Designed with low sodium ink to prevent contamination in cleanroom settings. Fisherbrand™ Cleanroom Pens come in black, blue, and red and feature HDPE construction.
$51.10 - $51.10
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
18-001-507
|
Fisherbrand™
18001507 |
Black |
Pack of 10 Each for $51.10
Case of 10 Pack for $511.00
|
|
|||||
Description
- Manufactured with very low sodium inks to prevent contamination
- Barrel and cap made from high density polyethylene (HDPE)
- Pens are packaged 10 per pack
- Available in Black (18-001-507), Blue (18-001-511), or Red (18-001-508)