Fisherbrand™ Cleanroom Vinyl Tape, 25.4 mm (1 in.)
This vinyl cleanroom tape is wound on a plastic core, can be written on with a cleanroom pen and is ideal for color coding, floor marking and light industrial use in clean construction and cleanroom support areas.
$565.62 - $1116.38
Specifications
Autoclavable Not Autoclavable
Adhesive Type Rubber
Material Vinyl
Sterility Non-sterile
Dimensions (LxW) 33 m x 25.4 mm (36 yd. x 1 in.)
View More Specs
Products 9
Catalog Number Mfr. No. Color Price Quantity  
Catalog Number Mfr. No. Color Price Quantity  
19120-3202
View Documents
Fisherbrand™
EGVFP1DB
Dark Blue
Case of 36 Each for $1,116.38
19120-3203
View Documents
Fisherbrand™
EGVFP1DG
Dark Green
Case of 36 Each for $575.26
19-120-3204
View Documents
Fisherbrand™
EGVFP1LB
Light Blue
Case of 36 Each for $575.01
19120-3205
View Documents
Fisherbrand™
EGVFP1LG
Light Green
Case of 36 Each for $565.62
19120-3206
View Documents
Fisherbrand™
EGVFP1OR
Orange
Case of 36 Each for $575.01
19-165-638
View Documents
Fisherbrand™
EGVFP1RE
Red
Case of 36 Each for $575.01
19120-3208
View Documents
Fisherbrand™
EGVFP1TR
Transparent
Case of 36 Each for $602.39
19-095-104
View Documents
Fisherbrand™
EGVFP1WH
White
Case of 36 Each for $588.35
19-165-639
View Documents
Fisherbrand™
EGVFP1YE
Yellow
Case of 36 Each for $588.35
Description
Description
For light industrial use, clean construction and cleanroom support
The Fisherbrand General Purpose Vinyl Cleanroom Tape forms a strong bond with plastics, polymers and stainless steel without leaving adhesive residue behind, when removed. Processed in a controlled environment, this tape is designed for light industrial use, clean construction and cleanroom support.
  • Conformable vinyl tape with rubber adhesive
  • Forms a strong bond with plastics, polymers and stainless steel
  • Does not leave adhesive residue when removed
  • Ideal for color coding, floor marking and general use in the cleanroom
  • Can be written on with cleanroom pen
  • Processed in a controlled environment
  • Wound on a plastic core and sleeve bagged
  • All rolls are 36 yd. long (33 m)
Specifications
Specifications
Not Autoclavable
Vinyl
33 m x 25.4 mm (36 yd. x 1 in.)
33 m
25.4 mm
Rubber
Non-sterile
36 yd.
1 in.
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'