Learn More
Fisherbrand™ Clear Boston Round Glass Bottles with White Polypropylene Caps
Clear Boston round Type III soda-lime glass is ideal for solvent, chemical or sample storage. Packaged in convenience packs with white polypropylene caps attached to maintain cleanliness
$45.92 - $388.32
Specifications
Color Clear
Material Glass (Type 3)
Closure Color White
Closure Included Yes
Mouth Narrow
View More Specs
Products 13
Catalog Number Mfr. No. Capacity (Metric) Closure Liner Quantity Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Closure Liner Quantity Price Quantity  
FB02911787
View Documents
N/A 60 mL PTFE/Polyethylene Foam 24
Case of 24 Each for $71.24
Only null left
FB02911790
View Documents
N/A 60 mL Pulp/Vinyl 288
Case of 288 Each for $388.32
Only null left
FB02911752
View Documents
N/A 60 mL Pulp/Vinyl 24
Case of 24 Each for $61.12
Only null left
FB02911780
View Documents
N/A 120 mL Pulp/Vinyl 24
Case of 24 Each for $91.80
Only null left
FB02911730
View Documents
N/A 120 mL Pulp/Vinyl 160
Case of 160 Each for $367.68
Only null left
FB02911788
View Documents
N/A 120 mL PTFE/Polyethylene Foam 24
Case of 24 Each for $106.32
Only null left
FB02911789
View Documents
N/A 240 mL PTFE/Polyethylene Foam 12
Case of 12 Each for $51.92
Only null left
FB02911732
View Documents
N/A 240 mL Pulp/Vinyl 108
Case of 108 Each for $347.00
Only null left
FB02911781
View Documents
N/A 240 mL Pulp/Vinyl 12
Case of 12 Each for $45.92
Only null left
FB02911753
missing translation for 'viewDocuments'
N/A 480 mL PTFE/Polyethylene Foam 12
Case of 12 Each for $79.20
Only null left
FB02911777
missing translation for 'viewDocuments'
N/A 480 mL Pulp/Vinyl 12
Case of 12 Each for $90.25
Only null left
FB02911778
missing translation for 'viewDocuments'
N/A 950 mL Pulp/Vinyl 12
Case of 12 Each for $97.84
Only null left
FB02911754
missing translation for 'viewDocuments'
N/A 950 mL PTFE/Polyethylene Foam 12
Case of 12 Each for $106.32
Only null left
Description
Description
  • General use bottles ideal for when content integrity and sample or product visibility is important
  • Narrow screw neck opening, curved shoulders, and a round body
  • Clear glass offers minimal chemical interference with contents and is appropriate when visibility is important but protection from UV rays is not necessary
  • Environmentally sensitive bottles help eliminate waste and help to insure product integrity for long term storage
Specifications
Specifications
Clear
White
Narrow
Polypropylene
Glass (Type 3)
Yes
Boston Round Bottle