Fisherbrand™ Clear Standard Wide Mouth Glass Bottles without Caps
Clear, Type III soda-lime glass ideal for general, small to large volume storage
 View more versions of this product
Catalog No. FB02911871
Description
Description
  • Features a wide mouth opening for easy transfer of contents
  • Applications include pharmaceutical, chemical, environmental, and food packaging and storing
  • Manufactured from clear flint glass
Specifications
Specifications
16 oz
16 oz.
Glass (Type 3)
63-400
473 mL
Clear
24
No