missing translation for 'onlineSavingsMsg'
Learn More

Clindamycin hydrochloride monohydrate

Clindamycin hydrochloride, CAS # 21462-39-5, is the salt of clindamycin, a derivative of the broad-spectrum bacteriostatic antibiotic lincomycin. | CAS: 58207-19-5 | C18H34Cl2N2O5S | 461.439 g/mol

$360.12 - $1,045.05

Chemical Identifiers

CAS 58207-19-5
Molecular Formula C18H34Cl2N2O5S
Molecular Weight (g/mol) 461.439
MDL Number MFCD07793327
InChI Key AUODDLQVRAJAJM-NFOLQJLNSA-N
PubChem CID 131632848
IUPAC Name (2S,4R)-N-[2-chloro-1-[(2S,3S,4S,5R,6R)-3,4,5-trihydroxy-6-methylsulfanyloxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide;hydrochloride
SMILES CCCC1CC(N(C1)C)C(=O)NC(C2C(C(C(C(O2)SC)O)O)O)C(C)Cl.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6140903
View Documents
Thermo Scientific Chemicals
J6140903
1 g N/A
Only null left
 
AAJ6140906
View Documents
Thermo Scientific Chemicals
J6140906
5 g N/A
Only null left
 
Description

Description

Clindamycin hydrochloride monohydrate is an antibacterial agent that is a semisynthetic analog of lincomycin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Clindamycin is a lincosamide antibiotic that disrupts protein synthesis by interfering with the aminoacyl-tRNA ribosome complex

• Clindamycin decreases sebaceous gland activity and blocks the neuromuscular junction

Application

• Clindamycin is commonly used in laboratory applications to inhibit the growth anaerobic microbes, but has some effectiveness against aerobic microbes as well

• It can be used in the laboratory study of cell culture for resistance testing, or in antibiotic allergic reaction studies

Specifications

Chemical Identifiers

58207-19-5
461.439
AUODDLQVRAJAJM-NFOLQJLNSA-N
(2S,4R)-N-[2-chloro-1-[(2S,3S,4S,5R,6R)-3,4,5-trihydroxy-6-methylsulfanyloxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide;hydrochloride
C18H34Cl2N2O5S
MFCD07793327
131632848
CCCC1CC(N(C1)C)C(=O)NC(C2C(C(C(C(O2)SC)O)O)O)C(C)Cl.Cl

Specifications

Clindamycin Hydrochloride Monohydrate
58207-19-5
1 g
MFCD07793327
14,2356
AUODDLQVRAJAJM-NFOLQJLNSA-N
(2S,4R)-N-[2-chloro-1-[(2S,3S,4S,5R,6R)-3,4,5-trihydroxy-6-methylsulfanyloxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide;hydrochloride
131632848
Clindamycin hydrochloride monohydrate
Powder
White
C18H34Cl2N2O5S
4070786
Soluble in water
CCCC1CC(N(C1)C)C(=O)NC(C2C(C(C(C(O2)SC)O)O)O)C(C)Cl.Cl
461.439
479.46 (461.44 Anhydrous)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 244-398-6

RTECSNumber : GF2275000

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only