missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clofazimine 98.0+%, TCI America™
$98.79 - $329.94
Chemical Identifiers
| CAS | 2030-63-9 |
|---|---|
| Molecular Formula | C27H22Cl2N4 |
| Molecular Weight (g/mol) | 473.401 |
| MDL Number | MFCD00056793 |
| InChI Key | WDQPAMHFFCXSNU-UHFFFAOYSA-N |
| Synonym | N,5-Bis(4-chlorophenyl)-3,5-dihydro-3-(isopropylimino)phenazin-2-amine |
| PubChem CID | 2794 |
| ChEBI | CHEBI:3749 |
| IUPAC Name | N,5-bis(4-chlorophenyl)-3-propan-2-yliminophenazin-2-amine |
| SMILES | CC(C)N=C1C=C2C(=NC3=CC=CC=C3N2C4=CC=C(C=C4)Cl)C=C1NC5=CC=C(C=C5)Cl |
Chemical Identifiers
| 2030-63-9 | |
| 473.401 | |
| WDQPAMHFFCXSNU-UHFFFAOYSA-N | |
| 2794 | |
| N,5-bis(4-chlorophenyl)-3-propan-2-yliminophenazin-2-amine |
| C27H22Cl2N4 | |
| MFCD00056793 | |
| N,5-Bis(4-chlorophenyl)-3,5-dihydro-3-(isopropylimino)phenazin-2-amine | |
| CHEBI:3749 | |
| CC(C)N=C1C=C2C(=NC3=CC=CC=C3N2C4=CC=C(C=C4)Cl)C=C1NC5=CC=C(C=C5)Cl |
Specifications
| 219°C | |
| Red-Yellow | |
| Crystal-Powder at 20°C | |
| MFCD00056793 | |
| WDQPAMHFFCXSNU-UHFFFAOYSA-N | |
| N,5-bis(4-chlorophenyl)-3-propan-2-yliminophenazin-2-amine | |
| 2794 | |
| 473.40 | |
| Clofazimine |
| 2030-63-9 | |
| 1 g | |
| C27H22Cl2N4 | |
| N,5-Bis(4-chlorophenyl)-3,5-dihydro-3-(isopropylimino)phenazin-2-amine | |
| CC(C)N=C1C=C2C(=NC3=CC=CC=C3N2C4=CC=C(C=C4)Cl)C=C1NC5=CC=C(C=C5)Cl | |
| 473.401 | |
| CHEBI:3749 | |
| ≥98.0% (HPLC,T) |
Safety and Handling
RTECSNumber : SG1578000
TSCA : No