Learn More
Clonidine hydrochloride, 98+%, Thermo Scientific Chemicals
$180.58 - $676.71
Chemical Identifiers
CAS | 4205-91-8 |
---|---|
Molecular Formula | C9H10Cl3N3 |
Molecular Weight (g/mol) | 266.55 |
MDL Number | MFCD00036705 |
InChI Key | ZNIFSRGNXRYGHF-UHFFFAOYSA-N |
Synonym | clonidine hydrochloride, clonidine hcl, catapres, 2-2,6-dichloroanilino-2-imidazoline hydrochloride, chlophazolin, dispaclonidin, normopresan, atensina, barclyd, capresin |
PubChem CID | 20179 |
IUPAC Name | N-(2,6-dichlorophenyl)-4,5-dihydro-1H-imidazol-2-amine;hydrochloride |
SMILES | C1CN=C(N1)NC2=C(C=CC=C2Cl)Cl.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6370703
|
Thermo Scientific Chemicals
J6370703 |
1 g |
Each for $180.58
|
|
|||||
AAJ6370706
|
Thermo Scientific Chemicals
J6370706 |
5 g |
Each for $676.71
|
|
|||||
Description
Clonidine hydrochloride tablets are indicated in the treatment of hypertension. It has found new uses, including treatment of some types of neuropathic pain, opioid detoxification, sleep hyperhidrosis, anaesthetic use, and off-label, to counter the side effects of stimulant medications such as methylphenidate or amphetamine. It is becoming a more accepted treatment for insomnia, as well as for relief of menopausal symptoms. Clonidine(4205-91-8) is increasingly used in conjunction with stimulants to treat attention-deficit hyperactivity disorder (ADHD).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in water (50 mg/ml), DMSO (75 mM), methanol, chloroform (slightly), and dehydrated alcohol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
4205-91-8 | |
266.55 | |
ZNIFSRGNXRYGHF-UHFFFAOYSA-N | |
20179 | |
C1CN=C(N1)NC2=C(C=CC=C2Cl)Cl.Cl |
C9H10Cl3N3 | |
MFCD00036705 | |
clonidine hydrochloride, clonidine hcl, catapres, 2-2,6-dichloroanilino-2-imidazoline hydrochloride, chlophazolin, dispaclonidin, normopresan, atensina, barclyd, capresin | |
N-(2,6-dichlorophenyl)-4,5-dihydro-1H-imidazol-2-amine;hydrochloride |
Specifications
4205-91-8 | |
C9H10Cl3N3 | |
1 g | |
4163525 | |
clonidine hydrochloride, clonidine hcl, catapres, 2-2,6-dichloroanilino-2-imidazoline hydrochloride, chlophazolin, dispaclonidin, normopresan, atensina, barclyd, capresin | |
ZNIFSRGNXRYGHF-UHFFFAOYSA-N | |
N-(2,6-dichlorophenyl)-4,5-dihydro-1H-imidazol-2-amine;hydrochloride | |
20179 | |
≥98% | |
Clonidine hydrochloride |
White | |
MFCD00036705 | |
UN2811 | |
14,2390 | |
Soluble in water (50mg/ml),DMSO (75 mM),methanol,chloroform (slightly),and dehydrated alcohol. | |
C1CN=C(N1)NC2=C(C=CC=C2Cl)Cl.Cl | |
266.55 | |
266.55 | |
Powder |
Safety and Handling
GHS H Statement
H301-H330
Toxic if swallowed.
Fatal if inhaled.
P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P330-P501c
H301-H330
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 224-121-5
RTECSNumber : NJ2490000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only