missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Clove Oil, FCC, 85%, Spectrum™ Chemical

CL130, 8000-34-8

Supplier:  Spectrum Chemical Mfg Cor CL130500MLGL

 View more versions of this product

Catalog No. 18601771


Explore available promotions

Description

Description

Spectrum™ Chemical Clove Oil, FCC is a flavor and fragarant ingredient. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Specifications

8000-34-8
Amber Glass Bottle
1.527-1.535
VEEIFXWJNCAVEQ-UHFFFAOYNA-N
methyl 2-amino-3-(1H-imidazol-5-yl)propanoate hydrochloride
85%
100%
C7H12ClN3O2
500 mL
Cl.COC(=O)C(N)CC1=CN=CN1
205.64
FCC

Chemical Identifiers

8000-34-8
205.64
methyl 2-amino-3-(1H-imidazol-5-yl)propanoate hydrochloride
C7H12ClN3O2
VEEIFXWJNCAVEQ-UHFFFAOYNA-N
Cl.COC(=O)C(N)CC1=CN=CN1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions