missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cloxacillin Sodium Salt Monohydrate 98.0+%, TCI America™
$93.21 - $268.43
Chemical Identifiers
| CAS | 7081-44-9 |
|---|---|
| Molecular Formula | C19H19ClN3NaO6S |
| Molecular Weight (g/mol) | 475.876 |
| MDL Number | MFCD00150735 |
| InChI Key | KCUWTKOTPIUBRI-VICXVTCVSA-M |
| Synonym | cloxacillin sodium, cloxapen, bactopen, cloxacillin sodium monohydrate, sodium cloxacillin monohydrate, tegopen, staphobristol-250, orbenin sodium hydrate, cloxacap, orbenin |
| PubChem CID | 23675743 |
| ChEBI | CHEBI:34978 |
| IUPAC Name | sodium;(2S,5R,6R)-6-[[3-(2-chlorophenyl)-5-methyl-1,2-oxazole-4-carbonyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate;hydrate |
| SMILES | CC1=C(C(=NO1)C2=CC=CC=C2Cl)C(=O)NC3C4N(C3=O)C(C(S4)(C)C)C(=O)[O-].O.[Na+] |
Chemical Identifiers
| 7081-44-9 | |
| 475.876 | |
| KCUWTKOTPIUBRI-VICXVTCVSA-M | |
| 23675743 | |
| sodium;(2S,5R,6R)-6-[[3-(2-chlorophenyl)-5-methyl-1,2-oxazole-4-carbonyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate;hydrate |
| C19H19ClN3NaO6S | |
| MFCD00150735 | |
| cloxacillin sodium, cloxapen, bactopen, cloxacillin sodium monohydrate, sodium cloxacillin monohydrate, tegopen, staphobristol-250, orbenin sodium hydrate, cloxacap, orbenin | |
| CHEBI:34978 | |
| CC1=C(C(=NO1)C2=CC=CC=C2Cl)C(=O)NC3C4N(C3=O)C(C(S4)(C)C)C(=O)[O-].O.[Na+] |
Specifications
| 7081-44-9 | |
| C19H19ClN3NaO6S | |
| 1 g | |
| KCUWTKOTPIUBRI-VICXVTCVSA-M | |
| sodium;(2S,5R,6R)-6-[[3-(2-chlorophenyl)-5-methyl-1,2-oxazole-4-carbonyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate;hydrate | |
| 23675743 | |
| 457.86 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD00150735 | |
| cloxacillin sodium, cloxapen, bactopen, cloxacillin sodium monohydrate, sodium cloxacillin monohydrate, tegopen, staphobristol-250, orbenin sodium hydrate, cloxacap, orbenin | |
| CC1=C(C(=NO1)C2=CC=CC=C2Cl)C(=O)NC3C4N(C3=O)C(C(S4)(C)C)C(=O)[O-].O.[Na+] | |
| 475.876 | |
| CHEBI:34978 | |
| ≥98.0% (HPLC) | |
| Cloxacillin Sodium Salt Monohydrate |
Safety and Handling
RTECSNumber : XH8750000
TSCA : Yes