Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers
Ensure optimal security for critical samples by using Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers.
$6277.34 - $8680.87
Catalog Number | Mfr. No. | Type | For Use With (Equipment) | Model | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | For Use With (Equipment) | Model | Price | Quantity | ||||
19-604-46
|
Thermo Scientific™
1960446 |
Field-Installed | TSE Series, Forma⢠7000 Series, HFU B and HFC Series, Revco™ CxF Series, TSC Series | CO2 Backup System |
Each for $7,312.53
|
|
||||
19-604-48
|
Thermo Scientific™
1960448 |
Field-Installed | TSC Series, HFU B and HFC Series, Revco™ CxF Series | LN2 Backup System |
Each for $6,770.06
|
|
||||
19-605-36
|
Thermo Scientific™
1960536 |
Field installed | TSE and TSD ULT Series | LN2 Backup System |
Each for $7,941.81
|
|
||||
FCO300A
|
Thermo Scientific™
FCO300A |
Field-Installed | 115V/60Hz models: Forma FDE30086, Revco RDE30086, HERAfreeze HDE30086, TDE30086 | CO2 Backup System |
Each for $7,432.98
|
|
||||
FCO300D
|
Thermo Scientific™
FCO300D |
Field-Installed | 208-230V/60Hz models: Forma FDE30086, Revco RDE30086, HERAfreeze HDE30086, TDE30086 | CO2 Backup System |
Each for $7,422.34
|
|
||||
FCO300V
|
Thermo Scientific™
FCO300V |
Field installed | 230V/50Hz models: Forma FDE30086, Revco RDE30086, HERAfreeze HDE30086, TDE30086 | CO2 Backup System |
Each for $7,422.34
|
|
||||
FCO300W
|
Thermo Scientific™
FCO300W |
Field-Installed | 230V/50Hz models (UK Plug): Forma FDE30086, Revco RDE30086, HERAfreeze HDE30086, TDE30086 | CO2 Backup System |
Each for $8,680.87
|
|
||||
FCO4567
|
Thermo Scientific™
FCO4567 |
Field-Installed | TSX Series, Forma 89000, HERAfreeze HLE, Revco RLE and TLE Series (400 box and above) | CO2 Backup System |
Each for $7,290.85
|
|
||||
FLN300A
|
Thermo Scientific™
FLN300A |
Field installed, standalone | 115V/60Hz models: Forma FDE30086, Revco RDE30086, HERAfreeze HDE30086, TDE30086 | LN2 Backup System |
Each for $7,269.15
|
|
||||
FLN300D
|
Thermo Scientific™
FLN300D |
Field installed, standalone | 208-230V/60Hz models: Forma FDE30086, Revco RDE30086, HERAfreeze HDE30086, TDE30086 | LN2 Backup System |
Each for $7,269.15
|
|
||||
FLN300V
|
Thermo Scientific™
FLN300V |
Field installed, standalone | 230V/50Hz models: Forma FDE30086, Revco RDE30086, HERAfreeze HDE30086, TDE30086 | LN2 Backup System |
Each for $7,269.15
|
|
||||
FLN300W
|
Thermo Scientific™
FLN300W |
Standalone | 230V/50Hz models (UK Plug): Forma FDE30086, Revco RDE30086, HERAfreeze HDE30086, TDE30086 | LN2 Backup System |
Each for $8,527.68
|
|
||||
FLN4567
|
Thermo Scientific™
FLN4567 |
Field-Installed | Revco™ UxF, HERAfreeze™ HFU-T, Thermo Scientific™ TSU Series | LN2 Backup System |
Each for $7,269.15
|
|
||||
FLN4567LE
|
Thermo Scientific™
FLN4567LE |
Field-installed | Thermo Scientific TSX Series | LN2 Backup System: maintains temperature down to -80°C with liquid nitrogen |
Each for $7,269.15
|
|
||||
FCO4567LE
|
Thermo Scientific™
FCO4567LE |
Field-installed | Thermo Scientific TSX Series | CO2 Backup System: maintains temperature down to -67°C with CO2 |
Each for $7,355.94
|
|
||||
FCO456LEA
|
Thermo Scientific™
FCO456LEA |
CO2 Backup System | Forma FDE, Revco RDE, HERAfreeze HDE, and Thermo Scientific TDE Series (400 box capacity and larger) | CO2 Backup System: maintains temperature down to -67°C with CO2 |
Each for $8,397.49
|
|
||||
FLN456LEA
|
Thermo Scientific™
FLN456LEA |
LN2 Backup System | Forma FDE, Revco RDE, HERAfreeze HDE, and Thermo Scientific TDE Series (400 box capacity and larger) | LN2 Backup System: maintains temperature down to -80°C with LN2 |
Each for $8,310.70
|
|
||||
332353G01
|
Thermo Scientific™
332353G01 |
Factory Installed | TDE Series Chest Freezers | LN2 Backup System |
Each for $6,734.53
|
|
||||
332353G02
|
Thermo Scientific™
332353G02 |
Factory Installed | TDE Series Chest Freezers | CO2 Backup System |
Each for $6,277.34
|
|
||||
332353G05
|
Thermo Scientific™
332353G05 |
Factory Installed | TDE Series Chest Freezers | LN2 Backup System |
Each for $6,734.53
|
|
||||
332353G06
|
Thermo Scientific™
332353G06 |
Factory Installed | TDE Series Chest Freezers | CO2 Backup System |
Each for $6,277.34
|
|
||||
LNTSXU
|
Thermo Scientific™
LNTSXU |
Factory-Installed | TSX Universal | LN2 Backup |
Each for $7,269.15
|
|
||||
FCOTSXU
|
Thermo Scientific™
FCOTSXU |
Field-installed | TSX Universal | CO2 Backup System |
Each for $8,614.47
|
|
||||
FLNTSXU
|
Thermo Scientific™
FLNTSXU |
Field-installed | TSX Universal | LN2 Backup System |
Each for $8,505.98
|
|
||||
Description
Ensure optimal security for critical samples by using Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers, for use with Thermo Scientific™ TSX, Forma™, HERAfreeze™, Revco™ and TS-Series freezers.
Features:- LN2 backup system maintains temperature down to -80°C with liquid nitrogen
- CO2 backup system maintains temperature down to -67°C with CO2
Ordering Information:
Available as field- or factory-installed. Models for 300 box capacity models are standalone. Field-installed units must be installed by qualified professionals.