missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ CO2 Resistant Shakers

Benefit from reliable around-the-clock operation ideally suited to keep your cells alive and flourishing within your working environment.

Supplier:  Thermo Scientific™ 88881101

 View more versions of this product

Catalog No. 88-881-101


Explore available promotions

Description

Description

Get reliable around-the-clock operation ideally suited to keep your cells alive and flourishing within your working environment with Thermo Scientific™ CO2 Resistant Shakers.

  • Orbital shaking through magnetic power eliminates the need to replace belts 
  • Small footprint ideal for use in a CO2 incubator 
  • Specially treated mechanical components protect up to 20% CO2 and 95% humidity conditions 
  • Convenient external control box with LED display allows for easy adjustments without opening the chamber door 
  • Minimal heat dissipation assures conditions within the CO2 chamber are not effected 
  • Stainless steel construction for protection from corrosive CO2 gassing conditions 
  • An extensive selection of accessories fits most applications 
  • CO2 resistant shaker can be used outside of CO2 incubator for maximum value
  • External control box for ease-of-use without disrupting your samples
A large selection of available accessories to fit most applications:
  • Wide variety of flask clamps available 
  • Fixed and adjustable tube racks available
  • 96-well microplate holder
  • Rubber and sticky mats 
  • Flask clamp starter kit for ease and flexibility
Applications:
  • Mammalian cell culture
  • Bacterial suspensions
  • Staining and destaining
  • Washing procedures
  • Hybridization
  • Solubility studies
  • Extraction procedures

Warranty:

  • 2 year Thermo Fisher Scientific standard limited warranty. See thermoscientific.com or your sales representative for details.
Specifications

Specifications

19 mm
±1 rpm
100/240 V, 50/60 Hz
4°C to 60°C
13 lb.
US Plug
50/60 Hz
29 lb.
CO2 Resistant Shaker
LED, speed and time
0 to 99:59 hr., Continuous
6 kg
30 to 300 rpm, ±1 rpm
100/240 V
CO2 Resistant Shaker
cCSAus, CE Mark, RoHS, WEEE
39.2°F to 140°F
11.8 x 14.2 x 3.8 in. (300 x 360 x 96 mm)
13 kg
CO2 Resistant Shaker
Shaker, control box, plugs
Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions