missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Greiner Bio-One Coagulation Tubes with Sodium Citrate Solution
For routine hemostasis testing
$36.14 - $36.14
Specifications
Product Type | Sodium Citrate Blood Collection Tube |
---|---|
Additive | 3.2% Citrate Solution |
Material | PET |
Tube Size | 13 x 75 mm |
Catalog Number | Mfr. No. | Additive | Closure Color | Capacity (Metric) | Product Type | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Additive | Closure Color | Capacity (Metric) | Product Type | Price | Quantity | ||
22-040-045
|
Greiner Bio-One
454322 |
3.2% Citrate Solution | Blue with White Ring | 1.8 mL | Sodium Citrate Blood Collection Tube |
Pack of 50 Each for $36.14
Case of 24 Pack for $867.36
|
|
||
22-040-119
|
Greiner Bio-One
454334 |
3.2% Citrate Solution | Blue with Black Ring | 2.7 mL | Sodium Citrate Blood Collection Tube |
Pack of 50 Each for $36.14
Case of 24 Pack for $867.36
|
|
||
Description
- 13 × 75mm tubes filled with buffered tri-sodium citrate solution
- 3.2 % (0.109 mol/L) concentration
- Mixing ratio is one part citrate to nine parts blood when tube is filled appropriately as indicated by fill mark on label
- Clear as glass with equal or better performance
- Molded to be virtually unbreakable, providing a better level of safety than glass
- 18-month shelf life from date of manufacture
- Blue nonscrew safety pull cap can be removed with a gentle pull
Specifications
Sodium Citrate Blood Collection Tube | |
3.2% Citrate Solution | |
PET | |
13 x 75 mm |