Learn More
Cobalt(II) acetate tetrahydrate, 98+%, Thermo Scientific Chemicals
$92.89 - $304.83
Chemical Identifiers
CAS | 6147-53-1 |
---|---|
Molecular Formula | C4H6CoO4·4H2O |
Molecular Weight (g/mol) | 249.08 |
MDL Number | MFCD00149569 |
InChI Key | ZBYYWKJVSFHYJL-UHFFFAOYSA-L |
Synonym | cobalt ii acetate tetrahydrate, cobalt acetate tetrahydrate, cobaltous acetate tetrahydrate, cobalt diacetate tetrahydrate, bis acetato tetraquacobalt, octan kobaltnaty czech, acetic acid, cobalt 2+ salt, tetrahydrate, octan kobaltnaty, ccris 9441 |
PubChem CID | 61372 |
IUPAC Name | cobalt(2+);diacetate;tetrahydrate |
SMILES | CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Co+2] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC293060500
|
Thermo Scientific Chemicals
293060500 |
50 g | Plastic bottle |
Each for $92.89
|
|
||||
AC293062500
|
Thermo Scientific Chemicals
293062500 |
250 g | Plastic bottle |
Each for $115.10
|
|
||||
AC293060010
|
Thermo Scientific Chemicals
293060010 |
1 kg | Plastic bottle |
Each for $304.83
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6147-53-1 | |
249.08 | |
ZBYYWKJVSFHYJL-UHFFFAOYSA-L | |
61372 | |
CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Co+2] |
C4H6CoO4·4H2O | |
MFCD00149569 | |
cobalt ii acetate tetrahydrate, cobalt acetate tetrahydrate, cobaltous acetate tetrahydrate, cobalt diacetate tetrahydrate, bis acetato tetraquacobalt, octan kobaltnaty czech, acetic acid, cobalt 2+ salt, tetrahydrate, octan kobaltnaty, ccris 9441 | |
cobalt(2+);diacetate;tetrahydrate |
Specifications
140.0°C | |
Pink to Pink-Red | |
50 g | |
C4H6CoO4·4H2O | |
MFCD00149569 | |
15, 2417 | |
Solubility in water: 380g/L (20°C). Other solubilities: soluble in acids, soluble in alcohols, soluble in pentyl acetate | |
CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Co+2] | |
249.08 | |
249.08 | |
Plastic bottle |
6147-53-1 | |
Crystalline Powder and/or Crystals | |
98% min. (Complexometry) | |
(CH3CO2)2Co·4H2O | |
04,99; 16,95 | |
cobalt ii acetate tetrahydrate, cobalt acetate tetrahydrate, cobaltous acetate tetrahydrate, cobalt diacetate tetrahydrate, bis acetato tetraquacobalt, octan kobaltnaty czech, acetic acid, cobalt 2+ salt, tetrahydrate, octan kobaltnaty, ccris 9441 | |
ZBYYWKJVSFHYJL-UHFFFAOYSA-L | |
cobalt(2+);diacetate;tetrahydrate | |
61372 | |
≥98% | |
Cobalt(II) acetate tetrahydrate, 97% |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Suspected of causing genetic defects.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer by inhalation.
May damage fertility.<br/
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Obtain speci
GHS Signal Word: Danger
RTECSNumber : AG3325000
RUO – Research Use Only