missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cobalt(II) acetate tetrahydrate, 98%, Thermo Scientific Chemicals

$52.04 - $176.60

Chemical Identifiers

CAS 6147-53-1
Molecular Formula C4H14CoO8
MDL Number MFCD00149569
InChI Key ZBYYWKJVSFHYJL-UHFFFAOYSA-L
Synonym cobalt ii acetate tetrahydrate, cobalt acetate tetrahydrate, cobaltous acetate tetrahydrate, cobalt diacetate tetrahydrate, bis acetato tetraquacobalt, octan kobaltnaty czech, acetic acid, cobalt 2+ salt, tetrahydrate, octan kobaltnaty, ccris 9441
PubChem CID 61372
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1837414
View Documents
Thermo Scientific Chemicals
A1837414
25 g
Each for $52.04
 
AAA1837422
View Documents
Thermo Scientific Chemicals
A1837422
100 g
Each for $65.21
 
AAA1837436
View Documents
Thermo Scientific Chemicals
A1837436
500 g
Each for $176.60
 
Description

Description

Cobalt(II) acetate tetrahydrate is used as an industrial catalyst to harden paints and varnishes, an active catalyst for oxidation and esterification reactions. It is also used to prepare metal complexes having unusual coordination geometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cobalt(II) acetate tetrahydrate is used as an industrial catalyst to harden paints and varnishes, an active catalyst for oxidation and esterification reactions. It is also used to prepare metal complexes having unusual coordination geometry.

Solubility
Soluble in water, alcohol, dilute acids and phenyl acetate.

Notes
Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong acids and bases.
Specifications

Chemical Identifiers

6147-53-1
MFCD00149569
cobalt ii acetate tetrahydrate, cobalt acetate tetrahydrate, cobaltous acetate tetrahydrate, cobalt diacetate tetrahydrate, bis acetato tetraquacobalt, octan kobaltnaty czech, acetic acid, cobalt 2+ salt, tetrahydrate, octan kobaltnaty, ccris 9441
C4H14CoO8
ZBYYWKJVSFHYJL-UHFFFAOYSA-L
61372

Specifications

6147-53-1
1.71
C4H14CoO8
Co(OOCCH3)2·4H2O
25 g
3731397
14,2433
Soluble in water,alcohol,dilute acids and phenyl acetate.
CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Co+2]
249.081
249.08 (177.02 Anhydrous)
Cobalt(II) acetate tetrahydrate
∼140°C (in -4H2O)
Odorless
1.542
MFCD00149569
UN3077
Hygroscopic
cobalt ii acetate tetrahydrate, cobalt acetate tetrahydrate, cobaltous acetate tetrahydrate, cobalt diacetate tetrahydrate, bis acetato tetraquacobalt, octan kobaltnaty czech, acetic acid, cobalt 2+ salt, tetrahydrate, octan kobaltnaty, ccris 9441
ZBYYWKJVSFHYJL-UHFFFAOYSA-L
cobalt(2+);diacetate;tetrahydrate
61372
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H350-H360-H341-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
May cause an allergic skin reaction.

P201-P202-P261-P271-P272-P280g-P281-P285-P302+P352-P304+P340-P308+P313-P333+P313-P342+P311-P363-P501c

H302-H317-H334-H335-H341-H350i-H360F

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 200-755-8

RTECSNumber : AG3325000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only