missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) carbonate hydrate

CAS: 57454-67-8 | CCoO3 | 118.94 g/mol

$105.71 - $927.81

Chemical Identifiers

CAS 57454-67-8
Molecular Formula CCoO3
Molecular Weight (g/mol) 118.94
MDL Number MFCD00149654
InChI Key ZOTKGJBKKKVBJZ-UHFFFAOYSA-L
Synonym cobalt ii carbonate hydrate, cobalt carbonate hydrate, acmc-20aqp2, co3.co.h2o, ksc169q7l, cobalt ii carbonate hydrate-co co min
PubChem CID 16211458
SMILES [Co++].[O-]C([O-])=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC192100500
SDS View Documents
Thermo Scientific Chemicals
192100500
50 g Plastic bottle N/A
Only null left
 
AC192102500
SDS View Documents
Thermo Scientific Chemicals
192102500
250 g Plastic bottle N/A
Only null left
 
AC192100010
SDS View Documents
Thermo Scientific Chemicals
192100010
1 kg Plastic bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

57454-67-8
118.94
ZOTKGJBKKKVBJZ-UHFFFAOYSA-L
16211458
CCoO3
MFCD00149654
cobalt ii carbonate hydrate, cobalt carbonate hydrate, acmc-20aqp2, co3.co.h2o, ksc169q7l, cobalt ii carbonate hydrate-co co min
[Co++].[O-]C([O-])=O

Specifications

57454-67-8
Mass, Crystals, Crystalline Powder and/or Chunks
CCoO3
MFCD00149654
cobalt ii carbonate hydrate, cobalt carbonate hydrate, acmc-20aqp2, co3.co.h2o, ksc169q7l, cobalt ii carbonate hydrate-co co min
ZOTKGJBKKKVBJZ-UHFFFAOYSA-L
λ²-cobalt(2+) carbonate
16211458
≥45.5% (Cobalt) (Complexometry)
Cobalt(II) carbonate hydrate
White to Yellow
50 g
CoCO3·xH2O
14,2436
Solubility in water: insoluble
[Co++].[O-]C([O-])=O
118.94
118.94
Plastic bottle
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of causing genetic defects.
Very toxic to aquatic life with long lasting effects.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer by inhalation.
May caus

GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
Wear p

GHS Signal Word: Danger

EINECSNumber : 208-169-4

RTECSNumber : FF9450050

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only