missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cobalt(II) citrate dihydrate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2408814

 View more versions of this product

Catalog No. AAB2408814


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cobalt(II) citrate dihydrate
6424-15-3
C12H20Co3O16
UN3077
SKMIIAIULKZNRQ-UHFFFAOYSA-N
cobalt;2-hydroxypropane-1,2,3-tricarboxylic acid;dihydrate
131882668
98%
∼150°C (decomposition)
25 g
MFCD02684007
cobalt ii citrate dihydrate
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O.O.[Co].[Co].[Co]
597.076
591.02 (554.99 Anhydrous)

Chemical Identifiers

6424-15-3
597.076
SKMIIAIULKZNRQ-UHFFFAOYSA-N
131882668
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O.O.[Co].[Co].[Co]
C12H20Co3O16
MFCD02684007
cobalt ii citrate dihydrate
cobalt;2-hydroxypropane-1,2,3-tricarboxylic acid;dihydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H350-H360-H341-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
May cause an allergic skin reaction.

P201-P202-P261-P264b-P270-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P341-P308+P313-P330-P333+P313-P342+P311-P363-P501c

H302-H317-H334-H341-H350-H360

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 212-751-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only