missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cobalt(II) hydroxide, 95%, pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 453890010

 View more versions of this product

Catalog No. AC453890010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Specifications

Cobalt(II) hydroxide
1 kg
CoH2O2
MFCD00016015
cobalt hydroxide, cobalt ii hydroxide, cobaltous hydrate, acmc-20aleg, cobalt 2+ dihydroxide, cobalt ii hydroxide metals basis
[OH-].[OH-].[Co++]
92.95
92.95
Pure
21041-93-0
95%
Co(OH)2
15,2426
ASKVAEGIVYSGNY-UHFFFAOYSA-L
cobalt(2+);dihydroxide
10129900
95%

Chemical Identifiers

21041-93-0
92.95
ASKVAEGIVYSGNY-UHFFFAOYSA-L
10129900
CoH2O2
MFCD00016015
cobalt hydroxide, cobalt ii hydroxide, cobaltous hydrate, acmc-20aleg, cobalt 2+ dihydroxide, cobalt ii hydroxide metals basis
[OH-].[OH-].[Co++]
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Very toxic to aq

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED:

GHS Signal Word: Danger

EINECSNumber : 244-166-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only