missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cobalt(II) nitrate hexahydrate, 99%, pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 213090050

 View more versions of this product

Catalog No. AC213090050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cobalt(II) nitrate hexahydrate
99%
Brown-Red to Pink
5 kg
CoH12N2O12
MFCD00149647
cobalt nitrate hexahydrate, cobalt ii nitrate hexahydrate, cobaltous nitrate hexahydrate, cobalt dinitrate hexahydrate, cobalt 2+ nitrate hexahydrate, cobaltous nitrate, hexahydrate, kobalt ii-nitrat-hexahydrat, nitric acid, cobalt 2+ salt, hexahydrate
QGUAJWGNOXCYJF-UHFFFAOYSA-N
λ²-cobalt(2+) hexahydrate dinitrate
24821
291.02
Pure
1.88
55.0°C
10026-22-9
Adhering Crystalline Powder
98.5% min. (Complexometry)
Co(NO3)2·6H2O
15, 2428
Solubility in water: 1340g/L. Other solubilities: soluble in ethanol and acetone, soluble in most common organic solvents
O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
291.03
CHEBI:86214
99%
Plastic bottle
1.8800g/mL

Chemical Identifiers

10026-22-9
291.03
QGUAJWGNOXCYJF-UHFFFAOYSA-N
24821
O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
CoH12N2O12
MFCD00149647
cobalt nitrate hexahydrate, cobalt ii nitrate hexahydrate, cobaltous nitrate hexahydrate, cobalt dinitrate hexahydrate, cobalt 2+ nitrate hexahydrate, cobaltous nitrate, hexahydrate, kobalt ii-nitrat-hexahydrat, nitric acid, cobalt 2+ salt, hexahydrate
CHEBI:86214
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May intensify fire; oxidizer.
Harmful if swallowed.
Harmful if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Causes serious ey

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remov

GHS Signal Word: Danger

RTECSNumber : QU7355500

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only