missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) phosphate, anhydrous, 98%

CAS: 13455-36-2 | Co3O8P2 | 366.739 g/mol

$113.70 - $337.18

Chemical Identifiers

CAS 13455-36-2
Molecular Formula Co3O8P2
Molecular Weight (g/mol) 366.739
MDL Number MFCD00016028
InChI Key ZBDSFTZNNQNSQM-UHFFFAOYSA-H
Synonym cobalt phosphate, cobaltous phosphate, cobalt ii phosphate, tricobalt bis orthophosphate, unii-0b5m38t47h, cobalt ii phosphate, anhydrous, phosphoric acid, cobalt 2+ salt 2:3, 3co.2po4, cobalt ii phosphate octahydrate, phosphoric acid,cobalt 2+ salt 2:3
PubChem CID 61615
IUPAC Name cobalt(2+);diphosphate
SMILES [O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Co+2].[Co+2].[Co+2]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2206614
View Documents
Thermo Scientific Chemicals
B2206614
25 g
Each for $113.70
Only null left
 
AAB2206622
View Documents
Thermo Scientific Chemicals
B2206622
100 g
Each for $337.18
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13455-36-2
366.739
ZBDSFTZNNQNSQM-UHFFFAOYSA-H
61615
[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Co+2].[Co+2].[Co+2]
Co3O8P2
MFCD00016028
cobalt phosphate, cobaltous phosphate, cobalt ii phosphate, tricobalt bis orthophosphate, unii-0b5m38t47h, cobalt ii phosphate, anhydrous, phosphoric acid, cobalt 2+ salt 2:3, 3co.2po4, cobalt ii phosphate octahydrate, phosphoric acid,cobalt 2+ salt 2:3
cobalt(2+);diphosphate

Specifications

13455-36-2
Co3O8P2
cobalt phosphate, cobaltous phosphate, cobalt ii phosphate, tricobalt bis orthophosphate, unii-0b5m38t47h, cobalt ii phosphate, anhydrous, phosphoric acid, cobalt 2+ salt 2:3, 3co.2po4, cobalt ii phosphate octahydrate, phosphoric acid,cobalt 2+ salt 2:3
[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Co+2].[Co+2].[Co+2]
366.739
366.74
Hygroscopic
25 g
MFCD00016028
ZBDSFTZNNQNSQM-UHFFFAOYSA-H
cobalt(2+);diphosphate
61615
98%
Cobalt(II) phosphate, anhydrous
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c

H302-H315-H319-H335-H350

EINECSNumber : 236-655-6

RTECSNumber : TB9279000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only