Learn More
Cobalt(III) 2,4-pentanedionate, Thermo Scientific Chemicals
$50.21 - $277.19
Chemical Identifiers
CAS | 21679-46-9 |
---|---|
Molecular Formula | C15H21CoO6 |
Molecular Weight (g/mol) | 356.26 |
MDL Number | MFCD00013488 |
InChI Key | VACIXPOMJJIDBM-UHFFFAOYSA-N |
Synonym | Cobalt(III) acetylacetonate |
IUPAC Name | 2,4,8,10,13,15-hexamethyl-1λ³,5λ³,7λ³,11λ³,12λ³,16λ³-hexaoxa-6-cobaltaspiro[5.5â¶.5â¶]hexadeca-1,4,7,10,12,15-hexaene-6,6,6-tris(ylium)-3,9,14-triide |
SMILES | CC1=[O][Co+3]234([O]=C(C)[CH-]1)[O]=C(C)[CH-]C(C)=[O]2.CC([CH-]C(C)=[O]3)=[O]4 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA1253606
|
Thermo Scientific Chemicals
01253606 |
5 g |
Each for $50.21
|
|
|||||
AA1253614
|
Thermo Scientific Chemicals
01253614 |
25 g |
Each for $113.14
|
|
|||||
AA1253622
|
Thermo Scientific Chemicals
01253622 |
100 g |
Each for $277.19
|
|
|||||
Description
Cobalt(III) 2,4-pentanedionate is used as a polymerization catalyst, vulcanizing agent, pigment in resins and glass. It is also used for the preparation of monoporphyrinates. Further, it is used in kinetic studies of iron-sulfur proteins. In addition to this, it reacts with trimethylaluminum to prepare Me(3Al/Co(acac)3.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
21679-46-9 | |
356.26 | |
VACIXPOMJJIDBM-UHFFFAOYSA-N | |
2,4,8,10,13,15-hexamethyl-1λ³,5λ³,7λ³,11λ³,12λ³,16λ³-hexaoxa-6-cobaltaspiro[5.5â¶.5â¶]hexadeca-1,4,7,10,12,15-hexaene-6,6,6-tris(ylium)-3,9,14-triide |
C15H21CoO6 | |
MFCD00013488 | |
Cobalt(III) acetylacetonate | |
CC1=[O][Co+3]234([O]=C(C)[CH-]1)[O]=C(C)[CH-]C(C)=[O]2.CC([CH-]C(C)=[O]3)=[O]4 |
Specifications
21679-46-9 | |
Light | |
MFCD00013488 | |
Hygroscopic; Moisture sensitive | |
Soluble in methanol and halogenated solvents. Slightly soluble in acetone,aromatic solvents and aliphatic solvents. Insoluble in water. | |
CC1=[O][Co+3]234([O]=C(C)[CH-]1)[O]=C(C)[CH-]C(C)=[O]2.CC([CH-]C(C)=[O]3)=[O]4 | |
356.26 | |
Powder |
211°C (decomposition) | |
C15H21CoO6 | |
5 g | |
Cobalt(III) acetylacetonate | |
VACIXPOMJJIDBM-UHFFFAOYSA-N | |
2,4,8,10,13,15-hexamethyl-1λ³,5λ³,7λ³,11λ³,12λ³,16λ³-hexaoxa-6-cobaltaspiro[5.5â¶.5â¶]hexadeca-1,4,7,10,12,15-hexaene-6,6,6-tris(ylium)-3,9,14-triide | |
356.26 | |
Cobalt(III) 2,4-pentanedionate |
Safety and Handling
GHS H Statement
H317
May cause an allergic skin reaction.
P201-P202-P261-P264b-P270-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P341-P308+P313-P330-P333+P313-P342+P311-P363-P501c
H302-H317-H334-H350
EINECSNumber : 244-527-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only