missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cobaltous Chloride Hexahydrate (Crystalline/Certified ACS), Fisher Chemical™
Supplier: Thermo Fisher Scientific FLC371100
Specifications
1048.9°C | |
86°C | |
Violet | |
Powder/Solid | |
98.0 to 102.0% | |
MFCD00149652 | |
Cobalt (II) Chloride | |
O.O.O.O.O.O.[Cl-].[Cl-].[Co++] | |
237.92 | |
98.0 to 102.0% | |
Pass Test | |
Negligible |
Cobaltous Chloride Hexahydrate | |
7791-13-1 | |
4.9 | |
100 g | |
Cl2CoH12O6 | |
UN 3288 | |
GFHNAMRJFCEERV-UHFFFAOYSA-L | |
λ²-cobalt(2+) hexahydrate dichloride | |
237.93 | |
Certified ACS | |
Glass Bottle |
Chemical Identifiers
7791-13-1 | |
237.92 | |
GFHNAMRJFCEERV-UHFFFAOYSA-L | |
λ²-cobalt(2+) hexahydrate dichloride |
Cl2CoH12O6 | |
MFCD00149652 | |
Cobalt (II) Chloride | |
O.O.O.O.O.O.[Cl-].[Cl-].[Co++] |
Safety and Handling
DANGER!
Emergency Overview
Possible cancer hazard. May cause cancer based on animal data. May cause cancer by inhalation. Harmful if swallowed. Irritating to eyes, respiratory system and skin. May cause allergic respiratory and skin reaction. May impair fertility. Possible risks of irreversible effects. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
NFPA
Health:3 Flammability:0 Instability:0
Signal Word: DANGER!
DOTInformation : DOT Class 6.1, : Poison