Lovibond™ COD LMR Tube Test, 15-300 mg/L
Each pack contains 25 COD vials, which are made of high-quality optical glass with a 16 mm diameter. Vials are packed in an EPS foam container which complies with global shipping and safety standards.
Supplier:  Lovibond™ 2423120
Catalog No. 01-670-221
Explore available promotions
Description
Description
  • Chemical Oxygen Demand (COD) is a common water quality parameter for municipal wastewater and industrial wastewater applications. It is used to quantify the amount of organics in a water sample and indicates the amount of oxygen that is available to be consumed by reactions with that solution.
  • As results of a COD test are easy to reproduce and are available in two hours, it is widely used as a process control parameter. COD values give a good indication of what is currently happening in a system and over time it can be used to make predictions to further optimize the treatment process.
  • Barcoded vial for easy method selection when used with Lovibond Spectrophotometers
  • Pre-filled tubes reduce sample preparation and handling
  • Part of the complete range of COD products from Lovibond™
Specifications
Specifications
Chemical Oxygen Demand (COD)
Industrial Wastewater, Environmental Compliance, Municipal Wastewater
24 and 1 reagent blank
15 to 300 mg/L
Dichromate / H2SO4
Tube Test / Vial Test
Lovibond Spectrophotometers and Colorimeters
25 Tubes Tests
Lovibond RD125 Reactor
Sample prep and 2 hour digestion required