Kent Scientific CODA™ Monitor Single Channel Noninvasive Blood Pressure System DFS Item
Allows to accurately measure mouse and rat blood pressure.
Supplier:  Kent Scientific CODAM1
Catalog No. 13005105
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Includes: RightTemp Temperature Monitor/Homeothermic Controller, mouse and rat temperature sensor, far Infrared warming pad, non-contact infrared thermometer.
Description
Description
  • Utilizes Volume Pressure Recording (VPR) sensor technology
  • Clinically validated
  • Provides 99% correlation with telemetry and direct blood pressure measurements
  • With RightTemp™ module
Specifications
Specifications
Accurately measure mouse and rat blood pressure
CODA Monitor Single Channel Noninvasive Blood Pressure System
RightTemp module