Fisherbrand™ Cold Water Faucet
Fisherbrand™ Cold Water Faucets are standard Laboratory cold water faucets.
 View more versions of this product
Catalog No. 06000396
Explore available promotions
Description
Description
  • Deck mounted
  • Chrome plated brass with 8 in. swivel gooseneck
  • Valves are self-contained units with replaceable stainless steel seats
Specifications
Specifications
Chrome-plated Brass
1 Ea.
29.1 cm
33.3 cm
Four-arm Handle
11.5 in.
13.1 in.