Andwin Scientific Cole-Parmer Instrument Co. Glas-Col™ Flexible Fabric Heating Mantles
Supplier: Andwin Scientific 3622540
Specifications
CSA | |
1000 mL | |
96.7 mm | |
380w | |
33.8 oz. | |
Commonly used method to heat a reaction mixture |
Electrical power | |
3.81 in. | |
115 V | |
450°C | |
842°F |