missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Colistin sulfate, Thermo Scientific Chemicals

Colistin Sulfate salt (2:5), CAS # 1264-72-8, (synonym: Polymyxin E Complex), is a complex mixture of antibiotics, with Colistin A and B being the primary components. It has anti-bacterial activity against Gram-positive and Gram-negative bacteria and may also be used as a neuromuscular blocker.

Supplier:  Thermo Scientific Chemicals J6091503

Catalog No. AAJ6091503


Explore available promotions

Description

Description

Colistin sulfate A Polymyxin potent antibiotic and apoptosis inducer. This compound induces apoptosis through interaction with the cytoplasmic membrane. Colistin is a key microbiological component in Colistin Oxolinic Acid Blood Agar utilized in the cultivation of Aminobacter aminovorans, Bacillus species, Hyphomicrobium species and Methylobacterium species. It is also a critical component is VCN Inhibitor & VCNT Inhibitor growth media used in the isolation of Neisseria species.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description
• Colistin Sulfate salt is a stable, cationic, water-soluble, white powder composed of over 30 components
• Originally isolated from B. polymyxa, it inhibits the growth of the Gram-negative (e.g. E. coli, P. aeruginosa, P. fluorescens, and S. enterica), and Gram-positive bacteria (e.g. L. lactis, P. polymyxa, P. acidilactici, and S. aureus) by permeabilizing the bacterial cell membrane
Applications
• Colistin Sulfate has potent anti-endotoxin activity rendered through its ability to bind lippopolysaccharides that make up bacterial endotoxins
• This compound is often used in the management of multi-drug resistant Gram-negative infections
Specifications

Specifications

Colistin Sulfate
Powder
White
MFCD00146495
14,2479
Soluble in water
Colistin sulfate
1264-72-8
1 g
UN2811
Polymyxin E
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P310-P330-P501c

H301

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 215-034-3

RTECSNumber : TR1500000

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only