missing translation for 'onlineSavingsMsg'
Learn More

Colloidal Silicon Dioxide, NF, EP, 99-100.5%, Spectrum™ Chemical

S1510, 7631-86-9, SiO2

$493.63 - $7088.80

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
IUPAC Name silanedione
SMILES O=[Si]=O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18601818
View Documents
Spectrum Chemical Mfg Cor
S1510100GM
100 g Plastic Bottle
Each for $493.63
Only null left
 
18601819
View Documents
Spectrum Chemical Mfg Cor
S1510500GM
500 g Poly Pail
Each for $1,468.00
Only null left
 
18601821
View Documents
Spectrum Chemical Mfg Cor
S15105KGBL
5 kg Fiber Drum
Each for $3,942.40
Only null left
 
18601820
View Documents
Spectrum Chemical Mfg Cor
S151010KGBL
10 kg Fiber Drum
Each for $7,088.80
Only null left
 
Description

Description

Spectrum™ Chemical Colloidal Silicon Dioxide, NF, EP is used both in the food and pharmaceutical industries as a powder flow helper in the production of tablets. All Spectrum Chemical NF, EP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Chemical Identifiers

7631-86-9
60.08
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
O=[Si]=O
O2Si
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
silanedione

Specifications

7631-86-9
Silicon Dioxide
O2Si
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
Plastic Bottle
O=[Si]=O
60.08
Multi-Compendial
100%
100 g
2.5%
5%
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
silanedione
99 to 100.5%
Colloidal Silicon Dioxide
Videos
SDS
Documents

Documents